EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Effect of some P-glycoprotein modulators on Rhodamine-123 absorption in guinea-pig ileum.
PMID 18524592
Abstract
Several reference compounds such as Cyclosporin A, Tamoxifen, Verapamil, and our compound 1, known as P-gp modulators, have been tested for their P-gp modulating activity in isolated organ bath. Compound 1 showed the best result in organ bath experiment (EC(50)=14.7 microM), Cyclosporin A and Tamoxifen displayed EC(50)=25.2 and 39.4 microM, respectively.