EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Expanding dialogues: from natural autoinducers to non-natural analogues that modulate quorum sensing in Gram-negative bacteria.
PMID 18568169
Abstract
Bacteria are capable of "communicating" their local population densities via a process termed quorum sensing (QS). Gram-negative bacteria use N-acylated l-homoserine lactones (AHLs), in conjunction with their cognate LuxR-type receptors, as their primary signalling circuit for QS. In this critical review, we examine AHL signalling in Gram-negative bacteria with a primary focus on the design of non-natural AHLs, their structure-activity relationships, and their application in chemical biological approaches to study QS (72 references).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
804231
2-bromo-N-[(3S)-tetrahydro-2-oxo-3-furanyl]-Benzeneacetamide
C12H12BrNO3