EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Cinnamoyl- and hydroxycinnamoyl amides of glaucine and their antioxidative and antiviral activities.
PMID 18590964
Abstract
The aporphine alkaloid glaucine has been converted into 3-aminomethylglaucine and its free amino group has been linked to cinnamic, ferulic, sinapic, o-, and p-coumaric acids. The antioxidative potential of the synthesized amides was studied against DPPH(*) test. All of the tested compounds demonstrated higher radical scavenging activity than glaucine and 3-aminomethylglaucine, and lower antioxidative effect than the free hydroxycinnamic acids. The newly synthesized compounds were tested in vitro for antiviral activity against viruses belonging to different taxonomic groups.