EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Inhibition of acetylcholinesterase, beta-amyloid aggregation, and NMDA receptors in Alzheimer's disease: a promising direction for the multi-target-directed ligands gold rush.
PMID 18605718
Abstract
Alzheimer's disease (AD) is a multifactorial syndrome with several target proteins contributing to its etiology. To confront AD, an innovative strategy is to design single chemical entities able to simultaneously modulate more than one target. Here, we present compounds that inhibit acetylcholinesterase and NMDA receptor activity. Furthermore, these compounds inhibit AChE-induced Abeta aggregation and display antioxidant properties, emerging as lead candidates for treating AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
543896 4-Hydroxycarbazole, 95%
C12H9NO