EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Formation of a liquid organic ion associate in aqueous solution and its application to the GF-AAS determination of trace cadmium in environmental water as a complex with 2-(5-bromo-2-pyridylazo)-5-(N-propyl-N-sulfopropylamino)phenol.
PMID 18614838
Abstract
The formation of a liquid organic ion associate in an aqueous sample was applied to the concentration and determination of cadmium in environmental water samples. Cadmium was converted into a complex with 2-(5-bromo-2-pyridylazo)-5-(N-propyl-N-sulfopropylamino)phenol (5-Br-PAPS) in a 40-mL sample solution, and was extracted into a liquid ion associate of phenolsulfonate and benzethonium during phase formation. More than 400-fold enrichment was easily attained by this technique, because the volume of the liquid organic phase formed was very small, ca. 2 microL. After dilution of the organic phase with a small volume of 2-methoxyethanol, the cadmium in the solution was determined by GF-AAS. The detection limit was 0.09 ng/L (3sigma(b)). This method was applied to the determination of cadmium in river water and seawater.