EMAIL THIS PAGE TO A FRIEND
Current opinion in critical care
Is there a role for sodium bicarbonate in treating lactic acidosis from shock?
PMID 18614899
Abstract
Bicarbonate therapy for severe lactic acidosis remains a controversial therapy. The most recent 2008 Surviving Sepsis guidelines strongly recommend against the use of bicarbonate in patients with pH at least 7.15, while deferring judgment in more severe acidemia. We review the mechanisms causing lactic acidosis in the critically ill and the scientific rationale behind treatment with bicarbonate. There is little rationale or evidence for the use of bicarbonate therapy for lactic acidosis due to shock. We agree with the Surviving Sepsis guidelines recommendation against the use of bicarbonate for lactic acidosis for pH at least 7.15 and we further recommend a lower target pH of 7.00 or less. If bicarbonate is used, consideration must be given to slow infusion and a plan for clearing the CO2 that is produced and measuring and correcting ionized calcium as the resultant 10% drop may decrease cardiac and vascular contractility and responsiveness to catecholamines. When continuous renal replacement therapy is used during severe acidosis, we recommend bicarbonate-based replacement fluid over citrate as citrate may increase the strong ion gap. Effective therapy of lactic acidosis due to shock is to reverse the cause.