EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
Tyrphostins as molecular tools and potential antiproliferative drugs.
PMID 1862529
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T182
Tyrphostin A9, solid
C18H22N2O