EMAIL THIS PAGE TO A FRIEND
Vascular health and risk management
Update on the use of fibrates: focus on bezafibrate.
PMID 18629356
Abstract
Low-density lipoprotein-cholesterol (LDL-C) is a well established coronary heart disease (CHD) risk factor. However, the ability of this metabolic risk factor alone to identify individuals at rigk for future CHD events is limited. The raised triglycerides-low high-density lipoprotein-cholesterol (HDL-C) dyslipidaemia was shown to be an important cardiovascular risk factor independently of LDL-C levels. Fibric acid derivatives (fibrates) have been used in clinical practice for more than 2 decades as a class of agents known to decrease triglyceride levels while substantially increasing HDL-C levels. Through peroxisome proliferator-activated alpha-receptors, fibrates have a significant impact on the synthesis of several apolipoproteins and enzymes of lipoprotein metabolism as well as on the expression of several genes involved in fibrinolysis and inflammation. Data from recent primary and secondary prevention clinical trials demonstrate the efficacy of fibrate therapy in patients with the raised triglycerides-low HDL-C dyslipidaemia. This review summarizes current data regarding mechanism of action and the metbolic effects of fibrates, as well as results from major clinical trials on the efficacy of this mode of lipid lowering therapy. In addition, recent data from subgroup analyses of the Bezafibrate Infarction Prevention trial, demonstrating several important metabolic and long-term cardiovascular effects of bezafibrate therapy, are detailed.