EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on borneol.
PMID 18640181
Abstract
A toxicologic and dermatologic review of borneol when used as a fragrance ingredient is presented.