EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on l-linalool.
PMID 18640207
Abstract
A toxicologic and dermatologic review of l-linalool when used as a fragrance ingredient is presented.
Related Materials