EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on isoborneol.
PMID 18640231
Abstract
A toxicologic and dermatologic review of isoborneol when used as a fragrance ingredient is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I13901
Isoborneol, 95%
C10H18O
W215805
Isoborneol, ≥95%, FG
C10H18O