EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on geranyl linalool.
PMID 18640234
Abstract
A toxicologic and dermatologic review of geranyl linalool when used as a fragrance ingredient is presented.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48809
Geranyllinalool, technical, ≥95% (GC)
C20H34O