EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on (-)-alpha-terpineol.
PMID 18644418
Abstract
A toxicologic and dermatologic review of p-menth-1-en-8-ol (S) when used as a fragrance ingredient is presented.