EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and structure-activity relationships of berberine analogues as a novel class of low-density-lipoprotein receptor up-regulators.
PMID 18644725
Abstract
Berberine (BBR, 1) is a novel cholesterol-lowering agent that up-regulates low-density-lipoprotein receptor (LDLR) expression through a mechanism different from that of statins. Because of the unique mode of action and good safety record, BBR provoked our interest to do structure modification at different domains for its cholesterol-lowering activity. Nineteen BBR analogues with substituents on the benzene ring D were synthesized in the present study. The analysis of structure-activity relationship (SAR) indicated that the two methoxyl groups in an ortho-distribution on this benzene ring afforded a good activity. Among the 19 analogues, compound 8j bearing a methoxyl at both 10- and 11-position showed an increased LDLR up-regulatory activity in respect to BBR, and therefore has been selected as a promising cholesterol-lowering drug candidate for further evaluation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3251
Berberine chloride form
C20H18ClNO4