EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Cyanopeptoline CB071: a cyclic depsipeptide isolated from the freshwater cyanobacterium Aphanocapsa sp.
PMID 18670126
Abstract
Cyanopeptolin CB071 (1), a trypsin inhibitor, was isolated from the freshwater cyanobacterium Aphanocapsa sp. Its complete structure was determined by detailed NMR spectroscopy and MS analyses, along with chemical reactions. The compound showed inhibition of trypsin at a concentration of IC50=2.5 microM.