EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Oleanolic acid and its derivatives: new inhibitor of protein tyrosine phosphatase 1B with cellular activities.
PMID 18707891
Abstract
Protein tyrosine phosphatase 1B is a key factor in the negative regulation of insulin pathway and a promising target for treatment of diabetes and obesity. Herein, a series of competitive inhibitors were optimized from oleanolic acid, a natural triterpenoid identified against PTP1B by screening libraries of traditional Chinese medicinal herbs. Modifying at 3 and 28 positions, we obtained compound 13 with a K(i) of 130 nM, which exhibited good selectivity between other phosphatases involved in insulin pathway except T-cell protein tyrosine phosphatase. Further evaluation in cell models illustrated that the derivatives enhanced insulin receptor phosphorylation in CHO/hIR cells and also stimulated glucose uptake in L6 myotubes with or addition of without insulin.