EMAIL THIS PAGE TO A FRIEND
Physical review letters
Highly conductive molecular junctions based on direct binding of benzene to platinum electrodes.
PMID 18764352
Abstract
Highly conductive molecular junctions were formed by direct binding of benzene molecules between two Pt electrodes. Measurements of conductance, isotopic shift in inelastic spectroscopy, and shot noise compared with calculations provide indications for a stable molecular junction where the benzene molecule is preserved intact and bonded to the Pt leads via carbon atoms. The junction has a conductance comparable to that for metallic atomic junctions (around 0.1-1G0), where the conductance and the number of transmission channels are controlled by the molecule's orientation at different interelectrode distances.