EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Cloning, expression, purification, crystallization and preliminary X-ray analysis of the human RuvBL1-RuvBL2 complex.
PMID 18765919
Abstract
The complex of RuvBL1 and its homologue RuvBL2, two evolutionarily highly conserved eukaryotic proteins belonging to the AAA(+) (ATPase associated with diverse cellular activities) family of ATPases, was co-expressed in Escherichia coli. For crystallization purposes, the flexible domains II of RuvBL1 and RuvBL2 were truncated. The truncated RuvBL1-RuvBL2 complex was crystallized using the hanging-drop vapour-diffusion method at 293 K. The crystals were hexagonal-shaped plates and belonged to either the orthorhombic space group C222(1), with unit-cell parameters a = 111.4, b = 188.0, c = 243.4 A and six monomers in the asymmetric unit, or the monoclinic space group P2(1), with unit-cell parameters a = 109.2, b = 243.4, c = 109.3 A, beta = 118.7 degrees and 12 monomers in the asymmetric unit. The crystal structure could be solved by molecular replacement in both possible space groups and the solutions obtained showed that the complex forms a dodecamer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3290
FLAG® Peptide, lyophilized powder
C41H60N10O20