EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
New lithocholic and chenodeoxycholic piperazinylcarboxamides with antiproliferative and pro-apoptotic effects on human cancer cell lines.
PMID 18768321
Abstract
Six new synthetic bile acid derivatives were synthesized and tested in vitro against various human cancer cells (glioblastoma multiforme (GBM), multiple myeloma (KMS-11), and colonic carcinoma (HCT-116) cell lines. The best activity was obtained with compound IIIb on multiple myeloma cells (LD(50): 8.5+/-0.5 microM). This activity was associated with Mcl-1 and PARP-1 cleavage, inhibition of NFkappaB signaling, and DNA fragmentation, demonstrating an apoptotic cell death signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
408166
1-(2-Pyridyl)piperazine, ≥99%
C9H13N3