EMAIL THIS PAGE TO A FRIEND
Clinical and experimental immunology
Leishmania donovani infection down-regulates TLR2-stimulated IL-12p40 and activates IL-10 in cells of macrophage/monocytic lineage by modulating MAPK pathways through a contact-dependent mechanism.
PMID 18778366
Abstract
The failure of Leishmania, an intracellular pathogen, to stimulate a pro-inflammatory response following entry into macrophages has been well reported. This occurs in spite of the fact that ligands for the toll-like receptors (TLR) have been recently shown on the parasite surface and their role in disease protection well documented. The outcome of infection in leishmaniasis is determined by the Th1 versus Th2 nature of the effector response and the generation of IL-12 and IL-10 by the infected macrophages is important for this decision. We evaluated the effect of L. donovani infection of monocytes (cell line THP-1, and monocytes derived from human peripheral blood) on Pam3cys (TLR2 ligand) and lipopolysaccharide (TLR4 ligand) stimulated production of IL-12p40 and IL-10. L. donovani infection caused suppression of TLR2 and TLR4-stimulated IL-12p40, with an increase in IL-10 production. Parasites also modulated the TLR2-stimulated mitogen-activated protein kinase (MAPK) pathway by suppressing MAPK P(38) phosphorylation and activating extracellular regulated kinase (ERK)1/2 phosphorylation. These effects could be reversed either by using a MAPK P(38) activator, anisomycin, or ERK1/2 inhibitor, U0126. L. donovani caused modulation of TLR2-stimulated MAPK pathways in a contact-dependent mechanism. In addition parasite structural integrity but not viability was required for suppression of TLR2-stimulated IL-12p40 and activation of IL-10. These observations suggest that L. donovani has evolved survival strategies that subvert the pro-inflammatory response generated through TLRs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8395
Filter membranes, nitrocellulose, MF membrane, pore size 0.22 μm, diam. 13 mm
N8520
Filter membranes, nitrocellulose, MF membrane, pore size 0.22 μm, diam. 25 mm
N8645
Filter membranes, nitrocellulose, MF membrane, pore size 0.22 μm, diam. 47 mm
N8770
Filter membranes, nitrocellulose, MF membrane, pore size 0.22 μm, diam. 142 mm
Z358657
Filter membranes, nitrocellulose, MF membrane, pore size 0.22 μm, diam. 293 mm
N8895
Filter membranes, nitrocellulose, MF membrane, pore size 0.45 μm, diam. 13 mm
N9020
Filter membranes, nitrocellulose, MF membrane, pore size 0.45 μm, diam. 25 mm
N9145
Filter membranes, nitrocellulose, MF membrane, pore size 0.45 μm, diam. 47 mm
N9270
Filter membranes, nitrocellulose, MF membrane, pore size 0.45 μm, diam. 142 mm
Z358649
Filter membranes, nitrocellulose, MF membrane, pore size 0.45 μm, diam. 293 mm
N9022
Filter membranes, nitrocellulose, MF membrane, pore size 0.65 μm, diam. 25 mm
N9147
Filter membranes, nitrocellulose, MF membrane, pore size 0.65 μm, diam. 47 mm
N8772
Filter membranes, nitrocellulose, MF membrane, pore size 0.8 μm, diam. 25 mm
N8897
Filter membranes, nitrocellulose, MF membrane, pore size 0.8 μm, diam. 47 mm
Z358738
Filter membranes, nitrocellulose, MF membrane, pore size 0.8 μm, diam. 293 mm
N3521
Filter membranes, nitrocellulose, MF membrane, pore size 5 μm, diam. 13 mm
N3896
Filter membranes, nitrocellulose, MF membrane, pore size 8 μm, diam. 13 mm
N4021
Filter membranes, nitrocellulose, MF membrane, pore size 8 μm, diam. 25 mm
N4146
Filter membranes, nitrocellulose, MF membrane, pore size 8 μm, diam. 47 mm
N4271
Filter membranes, nitrocellulose, MF membrane, pore size 8 μm, diam. 142 mm
N9395
Filter membranes, nitrocellulose, TF membrane, pore size 0.22 μm, diam. 13 mm
N9645
Filter membranes, nitrocellulose, TF membrane, pore size 0.22 μm, diam. 47 mm
N9770
Filter membranes, nitrocellulose, TF membrane, pore size 0.22 μm, diam. 142 mm
N9895
Filter membranes, nitrocellulose, TF membrane, pore size 0.45 μm, diam. 13 mm
N0146
Filter membranes, nitrocellulose, TF membrane, pore size 0.45 μm, diam. 25 mm
N0271
Filter membranes, nitrocellulose, TF membrane, pore size 0.45 μm, diam. 47 mm
N0521
Filter membranes, nitrocellulose, TF membrane, pore size 0.45 μm, diam. 142 mm
R5886 RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
R7388 RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
51536C RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture