EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Triterpenoids from the leaves of Diospyros kaki (persimmon) and their inhibitory effects on protein tyrosine phosphatase 1B.
PMID 18798681
Abstract
Phytochemical study on a methanol-soluble extract of the leaves of persimmon (Diospyros kaki) resulted in the isolation of two new ursane-type triterpenoids, 3alpha,19alpha-dihydroxyurs-12,20(30)-dien-24,28-dioic acid (1) and 3alpha,19alpha-dihydroxyurs-12-en-24,28-dioic acid (2), together with 12 known ursane- and oleanane-type triterpenoids (3-14). Triterpenoids with a 3beta-hydroxy group were found to inhibit protein tyrosine phosphatase 1B (PTP1B) activity, with IC50 values ranging from 3.1+/-0.2 to 18.8+/-1.3 microM, whereas those with a 3alpha-hydroxy moiety were not active.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89797
Ursolic acid, analytical standard
C30H48O3