EMAIL THIS PAGE TO A FRIEND
Journal of hypertension
Impaired angiotensin II-extracellular signal-regulated kinase signaling in failing human ventricular myocytes.
PMID 18806628
Abstract
Angiotensin II was reported to induce insulin-like growth factor-I and endothelin-1 gene expression and peptide release by ventricular cardiomyocytes. However, the progression from cardiac hypertrophy to failure in humans is characterized by a reduced myocyte expression of insulin-like growth factor-I and endothelin-1, notwithstanding the enhanced cardiac generation of angiotensin II. In the present study we investigated the functional status of the signaling pathways responsible for angiotensin II-induced endothelin-1 and insulin-like growth factor-I formation in human ventricular myocytes isolated from patients with dilated (n = 19) or ischemic (n = 14) cardiomyopathy and nonfailing donor hearts (n = 6).In human nonfailing ventricular myocytes, angiotensin II (100 nmol/l) induced insulin-like growth factor-I and endothelin-1 gene expression, and peptide release was mediated by extracellular signal-regulated kinase activation and inhibited by extracellular signal-regulated kinase antagonism (PD98059, 30 micromol/l), endothelin-1 formation being partially reduced also by c-Jun N-terminal kinase inhibition (SP600125, 10 micromol/l); insulin-like growth factor-I and endothelin-1 formations were unaffected by the inhibition of p38 mitogen-activated protein kinase (SB203580, 10 micromol/l) and Janus tyrosine kinase 2 (AG490, 10 micromol/l). In failing myocytes, angiotensin II failed to induce insulin-like growth factor-I and endothelin-1 formation; angiotensin II-induced extracellular signal-regulated kinase activation was significantly impaired (-88% vs. controls) although c-Jun NH2-terminal kinase activation was preserved. The impaired extracellular signal-regulated kinase phosphorylation in failing myocytes was associated with increased myocyte levels of mitogen-activated protein kinase phosphatases.Therefore, the altered growth factor production in failing myocytes is associated with a significant derangement in intracellular signaling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0625
Taurine, ≥99%
C2H7NO3S