EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Extractive spectrophotometric determination of TRODAT-1 hydrochloride in lyophilized kit.
PMID 18819514
Abstract
A simple, sensitive, and accurate spectrophotometric method has been developed for the assay of TRODAT-1 hydrochloride in lyophilized kit. The method is based on the formation of ion-pair association complex of TRODAT-1 with bromothymol blue (BTB) in disodium hydrogen phosphate/citric acid buffer of pH 4.0. The colored product was extracted with chloroform, and measured spectrophotometrically at 414 nm. Beer's law was obeyed in the range of 5-25 microg/ml with molar absorptivity of 2.75 x 10(4) l/mol/cm. Optimization of experimental conditions was described for the method. The proposed method has been successfully applied for the analysis of TRODAT-1 hydrochloride in lyophilized kit. No interference with pharmaceutical excipients was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
114413
Bromothymol Blue, ACS reagent, Dye content 95 %
C27H28Br2O5S
34656
Bromothymol Blue, pkg of 200 mL
C27H28Br2O5S