EMAIL THIS PAGE TO A FRIEND
Journal of the American Podiatric Medical Association
Gauze application of phenol for matrixectomy.
PMID 18820047
Abstract
Phenol matrixectomy is commonly used to treat onychocryptosis. The podiatric medical community has been progressively improving the technique of phenol application to avoid cases of burns. We describe a modification that uses gauze to provide a safe way for the phenol to be applied and prevents skin lesions due to phenol burns.