EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Preliminary SAR studies on non-apamin-displacing 4-(aminomethylaryl)pyrrazolopyrimidine K(Ca) channel blockers.
PMID 18824351
Abstract
An exploratory SAR study on a series of potent, non-apamin-displacing 4-(aminomethylaryl)pyrazolopyrimidine K(Ca) channel blockers is described and their selectivity against K(Ca) channel subtypes is reported. The most potent analog, 5-chloro-N-(thiophen-2-ylmethyl)pyrazolo[1,5-a]pyrimidin-7-amine (24) displayed sub-micromolar activity in both a thallium flux and whole-cell electrophysiology assay and did not displace apamin in a competitive binding study.