EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Use of the X-ray structure of the beta2-adrenergic receptor for drug discovery. Part 2: Identification of active compounds.
PMID 18829308
Abstract
The recently published X-ray structures of the beta(2)-adrenergic receptor are the first examples of ligand-mediated GPCR crystal structures. We have previously performed computational studies that examine the potential viability of these structures for use in drug design, exploiting known ligand activities. Our previous study and a newly reported beta(2)/Timolol X-ray complex provide validation of the computational approaches. In the present work, we use the X-ray structures to extract, via in silico high-throughput docking, compounds from proprietary and commercial databases and demonstrate the successful identification of active compounds by radioligand binding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53787
Carazolol, analytical standard
C18H22N2O2