EMAIL THIS PAGE TO A FRIEND
The Journal of law, medicine & ethics : a journal of the American Society of Law, Medicine & Ethics
Fixed-dose isosorbide dinitrate-hydralazine: race-based cardiovascular medicine benefit or mirage?
PMID 18840236
Abstract
Race is not a scientific category, but African Americans have increased prevalence and severity of heart failure. The African American Heart Failure trial showed the benefit of a combination of isosorbide dinitrate (a nitric oxide donor) and hydralazine (an antioxidant). Future research may unmask the reason for cardiovascular differences in therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0775000
Isosorbide dinitrate, European Pharmacopoeia (EP) Reference Standard
C6H8N2O8