EMAIL THIS PAGE TO A FRIEND
Journal of the Chemical Society
Synthetic antimalarials; some derivatives of phthalazine, quinoxaline, and isoquinoline.
PMID 18871271
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
368830
1,3-Dichloroisoquinoline, 97%
C9H5Cl2N