EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
A new synthesis of S-(beta-amino-beta-carboxyethyl) homocysteine (cystathionine) and S-bis(gamma-amino-gamma -carboxypropyl) sulfide (homolanthionine).
PMID 18902382
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3633
Cystathionine, ≥90% (HPLC)
C7H14N2O4S