EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
The design, synthesis and biological evaluation of 7-alkoxy-4-heteroarylamino-3-cyanoquinolines as dual inhibitors of c-Src and iNOS.
PMID 18930653
Abstract
Because both c-Src and iNOS are key regulatory enzymes in tumorigenesis, a new series of 4-heterocycle amine-3-quinolinecarbonitriles as potent dual inhibitors of both enzymes were designed, synthesized and evaluated as multiple targets agents in cancer therapy. All compounds were evaluated by two related enzyme inhibition assays and an anti-proliferation assay in vitro. The results showed that most compounds inhibited c-Src and iNOS well. The best compound 8 inhibited both enzymes with the IC(50) values of 34.8 nM and 26.7 microM. Several compounds also showed moderate anti-proliferation at 10 microM against colon and liver cancer cell lines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123129
2-Aminothiazole, 97%
C3H4N2S