EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Susceptibility of Mycobacterium bovis BCG vaccine strains to antituberculous antibiotics.
PMID 18955515
Abstract
Mycobacterium bovis BCG is one of the most commonly administered vaccines. Complications, including disseminated BCG disease, are rare but increasingly reported in immunodeficient children. There is growing recognition of the importance of differences between BCG vaccine strains. We determined the susceptibilities of five genetically distinct BCG vaccine strains to 12 antituberculous drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33703
Ofloxacin, VETRANAL, analytical standard
C18H20FN3O4
P7136
Pyrazinecarboxamide
C5H5N3O