EMAIL THIS PAGE TO A FRIEND
Talanta
Improved procedure for determining tellurium with Bismuthiol II.
PMID 18959997
Abstract
An improved procedure for the determination of tellurium with Bismuthiol II is described. Ammonium sulphate is found to increase the rate of complex formation and to enhance the sensitivity of the reaction by about 20%.