EMAIL THIS PAGE TO A FRIEND
Talanta
The synthesis of some lipophilic tetra-arylborates for use in membrane electrode preparation.
PMID 18965176
Abstract
Synthetic methods for preparing four lipophilic tetra-arylborates considered suitable candidates for use in membrane electrodes are described. Qualitative precipitation tests were performed with several cations. The sodium salts of the four tetra-arylborates were found to have limited stability but the cesium salts were relatively stable.