EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
The BH3-only protein Bik/Blk/Nbk inhibits nuclear translocation of activated ERK1/2 to mediate IFNgamma-induced cell death.
PMID 18981230
Abstract
IFNgamma induces cell death in epithelial cells, but the mediator for this death pathway has not been identified. In this study, we find that expression of Bik/Blk/Nbk is increased in human airway epithelial cells (AECs [HAECs]) in response to IFNgamma. Expression of Bik but not mutant BikL61G induces and loss of Bik suppresses IFNgamma-induced cell death in HAECs. IFNgamma treatment and Bik expression increase cathepsin B and D messenger RNA levels and reduce levels of phospho-extracellular regulated kinase 1/2 (ERK1/2) in the nuclei of bik(+/+) compared with bik(-/-) murine AECs. Bik but not BikL61G interacts with and suppresses nuclear translocation of phospho-ERK1/2, and suppression of ERK1/2 activation inhibits IFNgamma- and Bik-induced cell death. Furthermore, after prolonged exposure to allergen, hyperplastic epithelial cells persist longer, and nuclear phospho-ERK is more prevalent in airways of IFNgamma(-/-) or bik(-/-) compared with wild-type mice. These results demonstrate that IFNgamma requires Bik to suppress nuclear localization of phospho-ERK1/2 to channel cell death in AECs.