EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antiparasitic and antifungal evaluation of 2'-arylsubstituted-1H,1'H-[2,5']bisbenzimidazolyl-5-carboxamidines.
PMID 19010569
Abstract
A series of 2'-arylsubstituted-1H,1'H-[2,5']-bisbenzimidazolyl-5-carboxamidines were prepared in a six-step process starting from 4-amino-3-nitrobenzonitrile. The antiparasitic activity against Trypanosoma brucei rhodesiense (T.b.r.), Plasmodium falciparum (P.f.), Leishmania donovani (L.d.) and Trypanosoma cruzi (T.c.) and antifungal activity against Candida albicans and Candida krusei were evaluated in vitro. Several compounds showed promising in vitro activity against T.b.r., P.f. and C. albicans and had superior activity against P.f. as compared to chloroquine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4405
Podophyllotoxin
C22H22O8