EMAIL THIS PAGE TO A FRIEND
Arteriosclerosis, thrombosis, and vascular biology
Torcetrapib differentially modulates the biological activities of HDL2 and HDL3 particles in the reverse cholesterol transport pathway.
PMID 19038848
Abstract
Therapeutic strategies to raise low plasma HDL-cholesterol levels, with concomitant normalization of the intravascular metabolism, physicochemical properties, and antiatherogenic function of HDL particles, are a major focus in atherosclerosis prevention. Patients displaying Type IIB hyperlipidemia (n=14) and healthy controls (n=11) were recruited. After drug washout, dyslipidemic patients first received atorvastatin (10 mg/d) for 6 weeks and subsequently torcetrapib/atorvastatin (60/10 mg/d) for the same period. Partial CETP inhibition markedly reduced supranormal CE transfer rates to normal levels from HDL3 (-58%; P<0.0001) to apoB-lipoproteins; endogenous CE transfer rates from HDL2 to apoB-lipoproteins were markedly subnormal as compared to those in control subjects (10.7+/-0.9 versus 29.3+/-4.8 microg CE/h/mL plasma, respectively). Torcetrapib enhanced the subnormal capacity of HDL2 particles from dyslipidemic patients to mediate free cholesterol efflux via both SR-BI and ABCG1 pathways (+38%; P<0.003 and +35%; P<0.03, respectively) as compared to baseline. In vitro observations and in vivo studies in mice demonstrated that CETP inhibition was associated with an enhanced selective hepatic uptake of CE from HDL particles (1.7-fold; P<0.0003). CETP inhibition partially corrected the abnormal physicochemical and functional properties of HDL2 and HDL3 particles in type IIB hyperlipidemia. Enhanced hepatic selective uptake of HDL-CE may compensate for attenuated indirect CE transfer to apoB-containing lipoproteins via CETP attributable to torcetrapib.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0170
Torcetrapib, ≥98% (HPLC)
C26H25F9N2O4