EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Synthesis and cytotoxicity of bidesmosidic betulin and betulinic acid saponins.
PMID 19115839
Abstract
The naturally occurring cytotoxic saponin 28-O-beta-d-glucopyranosylbetulinic acid 3beta-O-alpha-l-arabinopyranoside (3) was easily synthesized along with seven bidesmosidic saponins starting from the lupane-type triterpenoids betulin (1) and betulinic acid (2). As highlighted by the preliminary cytotoxicity evaluation against A549, DLD-1, MCF7, and PC-3 human cancer cell lines, the bidesmosidic betulin saponin 22a, bearing alpha-l-rhamnopyranoside moieties at both C-3 and C-28 positions, was determined to be a potent cytotoxic agent (IC(50) 1.8-1.9 microM).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9757
Betulin, ≥98%
C30H50O2
855057
Betulinic acid, technical grade, 90%
C30H48O3