EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Oral and parenteral therapeutic options for outpatient urinary infections caused by enterobacteriaceae producing CTX-M extended-spectrum beta-lactamases.
PMID 19124661
Abstract
Effective therapeutic options are needed for community-onset urinary tract infections due to Escherichia coli strains that produce CTX-M extended-spectrum beta-lactamases. We examined 46 urinary isolates producing CTX-M against several oral or long-acting parenteral antimicrobial agents. Approximately 90% were susceptible to fosfomycin and to a combination of cefdinir plus amoxicillin-clavulanate. All were susceptible to ertapenem.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7118 Cefdinir, ≥95% (HPLC)
C14H13N5O5S2
46502
Nitrofurantoin, VETRANAL, analytical standard
C8H6N4O5