EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Carbohydrate binding specificity of recombinant human macrophage beta-glucan receptor dectin-1.
PMID 19129647
Abstract
Human macrophage dectin-1, a type II transmembrane beta-glucan receptor, was expressed as a fusion protein with an N-terminal hexahistidine tag and glutathione S-transferase in an Escherichia coli cell-free translation system, and assayed for binding specificity. Recombinant dectin-1 specifically bound to some beta-glucans, but not to other carbohydrates. The beta-glucan binding of recombinant dectin-1 was inhibited by laminarin, a soluble beta-glucan, and by laminarioligosaccharides, but not by other carbohydrates. These results suggest that recombinant human dectin-1 can be used as a useful probe in identifying ligands in humans and tonic foods due to its strict binding specificity.
Related Materials