EMAIL THIS PAGE TO A FRIEND
Cell death and differentiation
TGF-beta induces apoptosis in human B cells by transcriptional regulation of BIK and BCL-XL.
PMID 19136942
Abstract
Transforming growth factor-beta (TGF-beta) potently induces apoptosis in Burkitt's lymphoma (BL) cell lines and in explanted primary human B lymphocytes. The physiological relevance and mechanism of TGF-beta-mediated apoptosis induction in these cells remains to be determined. Here we demonstrate the requirement for TGF-beta-mediated regulation of BIK and BCL-X(L) to activate an intrinsic apoptotic pathway in centroblastic BL cells. TGF-beta directly induced transcription of BIK and a consensus Smad-binding element identified in the BIK promoter recruits TGF-beta-activated Smad transcription factor complexes in vivo. TGF-beta also transcriptionally repressed expression of the apoptosis inhibitor BCL-X(L). Inhibition of BCL-X(L) sensitised BL cells to TGF-beta-induced apoptosis whereas overexpression of BCL-X(L) or suppression of BIK by shRNA, diminished TGF-beta-induced apoptosis. BIK and BCL-X(L) were also identified as TGF-beta target genes in purified normal human centroblast B cells and immunohistochemical analyses of tonsil tissue revealed widespread TGF-beta receptor-regulated Smad activation and a focal pattern of BIK expression. Furthermore, using a selective inhibitor of the TGF-beta receptor we provide evidence that autocrine TGF-beta signalling through ALK5 contributes to the default apoptotic programme in normal human centroblasts undergoing spontaneous apoptosis. Our data suggests that TGF-beta may act as a physiological mediator of human germinal centre homoeostasis by regulation of BIK and BCL-X(L).
Related Materials