EMAIL THIS PAGE TO A FRIEND
Drug and therapeutics bulletin
Acipimox--a nicotinic acid analogue for hyperlipidaemia.
PMID 1914867
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7856 Acipimox, ≥99% (TLC)
C6H6N2O3