EMAIL THIS PAGE TO A FRIEND
Recent patents on anti-cancer drug discovery
The anti-cancer activity of noscapine: a review.
PMID 19149691
Abstract
Noscapine is an isoqiunoline alkaloid found in opium latex. Unlike most other alkaloids obtained from opium latex, noscapine is not sedative and has been used as antitussive drug in various countries. Recently, it has been introduced as an anti-mitotic agent. This drug can be used orally. When the resistance to other anti-cancer drugs such as paclitaxel manifests, noscapine might be effective. Therefore, noscapine and its analogs have great potential as novel anti-cancer agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
363960
(S,R)-Noscapine, 97%
C22H23NO7
N1300000
Noscapine, European Pharmacopoeia (EP) Reference Standard
C22H23NO7