EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Facile labeling of lipoglycans with quantum dots.
PMID 19150336
Abstract
Bacterial endotoxins or lipopolysaccharides (LPS) are among the most potent activators of the innate immune system, yet mechanisms of their action and in particular the role of glycans remain elusive. Efficient non-invasive labeling strategies are necessary for studying interactions of LPS glycans with biological systems. Here we report a new method for labeling LPS and other lipoglycans with luminescent quantum dots. The labeling is achieved by partitioning of hydrophobic quantum dots into the core of various LPS aggregates without disturbing the native LPS structure. The biofunctionality of the LPS-Qdot conjugates is demonstrated by the labeling of mouse monocytes. This simple method should find broad applicability in studies concerned with visualization of LPS biodistribution and identification of LPS binding agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3665 Lipopolysaccharides from Escherichia coli O111:B4, FITC conjugate