EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Occurrence of efflux mechanism and cephalosporinase variant in a population of Enterobacter aerogenes and Klebsiella pneumoniae isolates producing extended-spectrum beta-lactamases.
PMID 19164140
Abstract
We investigated the occurrence of multidrug resistance in 44 Enterobacter aerogenes and Klebsiella pneumoniae clinical isolates. Efflux was involved in resistance in E. aerogenes isolates more frequently than in K. pneumoniae isolates (100 versus 38% of isolates) and was associated with the expression of phenylalanine arginine beta-naphthylamide-susceptible active efflux. AcrA-TolC overproduction in E. aerogenes isolates was noted. An analysis of four E. aerogenes isolates for which cefepime MICs were high revealed no modification in porin expression but a new specific mutation in the AmpC beta-lactamase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56968
Sparfloxacin, ≥98.0% (HPLC)
C19H22F2N4O3