EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Targeted deletion of hepatocyte Ikkbeta confers growth advantages.
PMID 19171122
Abstract
Mice lacking hepatocyte IKKbeta (Ikkbeta(Delta hep)) are defective in TNFalpha-activation of hepatocellular transcription factor NF-kappaB, and highly susceptible to hepatotoxicity. Following diethylnitrosamine (DEN) exposure, Ikkbeta(Delta hep) mice develop more hepatocellular carcinoma (HCC) than control mice due partly to enhanced DEN-induced hepatocyte death. Here we show that Ikkbeta(Delta hep) hepatocytes display growth advantages over normal hepatocytes consisting of precocious PCNA and cyclin D1 expression during liver regeneration (shortened hepatocyte G(0)-->G(1) transitions), and enhanced recovery efficiency, cyclin D1 expression and cell proliferation after plating. Ex vivo deletion of Ikkbeta also accelerates hepatocyte growth. Ikkbeta(Delta hep) hepatocyte proliferative responses show heightened sensitivity to TGFalpha and TNFalpha, and heightened expression of fibronectin, collagens I/III, nidogen, beta-actin and integrin beta1 mRNAs. These findings suggest that altered mitogen signaling and expression of extracellular matrix and its associated components underlie growth advantages. Increased HCC development in Ikkbeta(Delta hep) mice may also be caused by growth advantages of surviving Ikkbeta-deleted hepatocytes.