EMAIL THIS PAGE TO A FRIEND
Organic letters
Novel approach to 3,4-dihydro-2(1H)-quinolinone derivatives via cyclopropane ring expansion.
PMID 19193029
Abstract
N-(1'-Alkoxy)cyclopropyl-2-haloanilines are transformed to 3,4-dihydro-2((1)H)-quinolinones via palladium-catalyzed cyclopropane ring expansion. The reaction tolerates a variety of functional groups such as ester, nitrile, ether, and ketone groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
415936
3,4-Dihydro-2(1H)-quinolinone, 98%
C9H9NO