EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Identification of stable S-adenosylmethionine (SAM) analogues derivatised with bioorthogonal tags: effect of ligands on the affinity of the E. coli methionine repressor, MetJ, for its operator DNA.
PMID 19194573
Abstract
The efficient synthesis of a range of stable SAM mimetics, and their ability to promote the binding of the E. coli methionine repressor (MetJ) to its operator DNA, is described.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462152
Methyl 4-iodobutyrate, 95%
C5H9IO2