EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and antibacterial activity evaluation of metronidazole-triazole conjugates.
PMID 19195884
Abstract
Synthesis and antibacterial activity of metronidazole-triazole conjugates are reported. Total 21 hybrid compounds have been synthesized with different substitution pattern on the triazole ring in order to study their influence on the antibacterial activity. These compounds demonstrated potent to weak antibacterial activity against Gram-positive, and Gram-negative bacteria. Six compounds have shown equal or better antibacterial activity against Gram-negative strains than the reference compound.