EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Ionothermal synthesis of zirconium phosphates and their catalytic behavior in the selective oxidation of cyclohexane.
PMID 19199313
Abstract
On their best behavior: Three zirconium compounds with one-, two-, and three-dimensional structures have been successfully synthesized by the ionothermal approach. The 3D zirconium phosphate (see picture; F green, H white, O red, P pink, Zr yellow) exhibits high catalytic performance, with a cyclohexane conversion ratio of 32% and cyclohexanone selectivity of up to 83%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
02482
Cyclohexanone, analytical standard
C6H10O