EMAIL THIS PAGE TO A FRIEND
ACS nano
TiO2-graphene nanocomposites. UV-assisted photocatalytic reduction of graphene oxide.
PMID 19206319
Abstract
Graphene oxide suspended in ethanol undergoes reduction as it accepts electrons from UV-irradiated TiO(2) suspensions. The reduction is accompanied by changes in the absorption of the graphene oxide, as the color of the suspension shifts from brown to black. The direct interaction between TiO(2) particles and graphene sheets hinders the collapse of exfoliated sheets of graphene. Solid films cast on a borosilicate glass gap separated by gold-sputtered terminations show an order of magnitude decrease in lateral resistance following reduction with the TiO(2) photocatalyst. The photocatalytic methodology not only provides an on-demand UV-assisted reduction technique but also opens up new ways to obtain photoactive graphene-semiconductor composites.
Related Materials